Product Details

SNP ID
rs147807256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4646144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTACCGGCACCTCCTCCTTCCAG[C/T]AGGGGAACAGTTTTTGCACCCAGGC
Phenotype
MIM: 615869
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFAIP8L1 PubMed Links

Gene Details

Gene
TNFAIP8L1
Gene Name
TNF alpha induced protein 8 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167942.1 Intron NP_001161414.1
NM_152362.2 Intron NP_689575.2
XM_005259487.3 Intron XP_005259544.1
XM_011527680.2 Intron XP_011525982.1

View Full Product Details