Product Details

SNP ID
rs148708853
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14054480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGTCTCCTCGCCTCCTCCCGTCT[G/T]GGCCCCCTCGGCCCCCAGCGGGCTT
Phenotype
MIM: 605350
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL27RA PubMed Links

Gene Details

Gene
IL27RA
Gene Name
interleukin 27 receptor subunit alpha
There are no transcripts associated with this gene.

Gene
PALM3
Gene Name
paralemmin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145028.1 1147 Missense Mutation AAG,CAG K383Q NP_001138500.1
XM_005259891.4 1147 Missense Mutation AAG,CAG K398Q XP_005259948.1
XM_011527965.2 1147 Missense Mutation AAG,CAG K371Q XP_011526267.1

View Full Product Details