Product Details

SNP ID
rs149477691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10256773 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCCGGAAGCCTTGGCCGCAGAA[A/G]GGCACTGGGCGGGCCCGGCATGGCA
Phenotype
MIM: 611823
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPL4 PubMed Links

Gene Details

Gene
MRPL4
Gene Name
mitochondrial ribosomal protein L4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015956.2 547 Silent Mutation AAA,AAG K131K NP_057040.2
NM_146387.1 547 Silent Mutation AAA,AAG K131K NP_666499.1
NM_146388.1 547 Silent Mutation AAA,AAG K131K NP_666500.1
XM_011528045.2 547 Silent Mutation AAA,AAG K131K XP_011526347.1

View Full Product Details