Product Details

SNP ID
rs149496880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11298884 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTCTTTTAGTGCATCCTGTCA[A/G]TGGTTATTGTCCTCATCATGGAAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPC102 PubMed Links

Gene Details

Gene
HSPC102
Gene Name
uncharacterized HSPC102
There are no transcripts associated with this gene.

Gene
TSPAN16
Gene Name
tetraspanin 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282509.1 370 Missense Mutation ATG,GTG M94V NP_001269438.1
NM_001282510.1 370 Intron NP_001269439.1
NM_012466.3 370 Missense Mutation ATG,GTG M94V NP_036598.1
XM_011527901.1 370 Missense Mutation ATG,GTG M94V XP_011526203.1
XM_011527902.1 370 Missense Mutation ATG,GTG M94V XP_011526204.1
XM_011527903.2 370 Missense Mutation ATG,GTG M31V XP_011526205.1

View Full Product Details