Product Details

SNP ID
rs150607510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41444073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCCTGGGGCTGGAGATGGGGG[C/T]GCTGCGGGAGGAACTGGTCACCATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5SL PubMed Links

Gene Details

Gene
ATP5SL
Gene Name
ATP5S like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167867.1 254 Intron NP_001161339.1
NM_001167868.1 254 Intron NP_001161340.1
NM_001167869.1 254 Intron NP_001161341.1
NM_001167870.1 254 Intron NP_001161342.1
NM_001167871.1 254 Intron NP_001161343.1
NM_001320838.1 254 Intron NP_001307767.1
NM_001320839.1 254 Intron NP_001307768.1
NM_001320840.1 254 Intron NP_001307769.1
NM_001320841.1 254 Intron NP_001307770.1
NM_001320844.1 254 Intron NP_001307773.1
NM_018035.2 254 Intron NP_060505.2
XM_011527065.2 254 Intron XP_011525367.1
Gene
ERICH4
Gene Name
glutamate rich 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130514.1 254 Missense Mutation GCG,GTG A81V NP_001123986.1
XM_017026119.1 254 Missense Mutation GCG,GTG A102V XP_016881608.1

View Full Product Details