Product Details

SNP ID
rs150679552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11507799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGGGCCCTCAACAAAGACAGGC[C/T]GGGCTGGATTGTGGCGGGCCAGGGC
Phenotype
MIM: 608388 MIM: 611371
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ECSIT PubMed Links

Gene Details

Gene
ECSIT
Gene Name
ECSIT signalling integrator
There are no transcripts associated with this gene.

Gene
ZNF653
Gene Name
zinc finger protein 653
There are no transcripts associated with this gene.

View Full Product Details