Product Details

SNP ID
rs150811577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38870947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATGGTCCACTTCCGGGCTGAGC[A/G]GGCCTTTCACCCAGGAGACCCCGTG
Phenotype
MIM: 604480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RINL PubMed Links

Gene Details

Gene
RINL
Gene Name
Ras and Rab interactor like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195833.1 728 Missense Mutation CCG,CTG P216L NP_001182762.1
NM_198445.3 728 Intron NP_940847.1
XM_006723015.2 728 Missense Mutation CCG,CTG P216L XP_006723078.1
XM_006723016.2 728 Missense Mutation CCG,CTG P102L XP_006723079.1
XM_011526454.2 728 Missense Mutation CCG,CTG P216L XP_011524756.1
XM_011526455.2 728 Missense Mutation CCG,CTG P216L XP_011524757.1
Gene
SIRT2
Gene Name
sirtuin 2
There are no transcripts associated with this gene.

View Full Product Details