Product Details

SNP ID
rs151262762
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16198047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCATGTCCGCCAGCGCCGTCTA[C/T]GTGCTGGACCTGAAGGGCAAGGTAC
Phenotype
MIM: 603535 MIM: 614554
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AP1M1 PubMed Links

Gene Details

Gene
AP1M1
Gene Name
adaptor related protein complex 1 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130524.1 194 Silent Mutation TAC,TAT Y7Y NP_001123996.1
NM_032493.3 194 Silent Mutation TAC,TAT Y7Y NP_115882.1
XM_017027420.1 194 Intron XP_016882909.1
XM_017027421.1 194 Intron XP_016882910.1
Gene
FAM32A
Gene Name
family with sequence similarity 32 member A
There are no transcripts associated with this gene.

View Full Product Details