Product Details

SNP ID
rs111998622
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:8013531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTGCTTTGCTGCTGACATACTT[G/T]GGATCAGGGGCAAAGCTCTGTGTCG
Phenotype
MIM: 608069
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERRFI1 PubMed Links

Gene Details

Gene
ERRFI1
Gene Name
ERBB receptor feedback inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018948.3 1331 Silent Mutation CCA,CCC P356P NP_061821.1
XM_005263477.2 1331 Silent Mutation CCA,CCC P305P XP_005263534.1
XM_006710697.2 1331 Silent Mutation CCA,CCC P356P XP_006710760.1
XM_011541596.2 1331 Silent Mutation CCA,CCC P281P XP_011539898.1

View Full Product Details