Product Details

SNP ID
rs112182116
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:51908481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAAGGTGACATCTCTCTGTGTGG[A/C]CACAGAGATAGATCATCTGCTCCTG
Phenotype
MIM: 179510
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RAB3B PubMed Links

Gene Details

Gene
RAB3B
Gene Name
RAB3B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002867.3 12319 UTR 3 NP_002858.2
XM_017001958.1 12319 Intron XP_016857447.1

View Full Product Details