Product Details

SNP ID
rs113255684
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:231163191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGAGCACGACGCTGCGGCTGG[C/T]CCGGCCTGCGGCGGTGCAGGCGGGA
Phenotype
MIM: 610584
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM67 PubMed Links

Gene Details

Gene
TRIM67
Gene Name
tripartite motif containing 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004342.3 264 Silent Mutation GGC,GGT G74G NP_001004342.3
NM_001300889.1 264 Intron NP_001287818.1
XM_011544192.2 264 Silent Mutation GGC,GGT G74G XP_011542494.1
XM_017001323.1 264 Silent Mutation GGC,GGT G74G XP_016856812.1

View Full Product Details