Product Details

SNP ID
rs114534381
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:158291362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTTTCTACCTCTTTGGATTAACTC[A/G]GGAGATTCAAGACCATGCAAGTCAA
Phenotype
MIM: 188360 MIM: 188340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD1B PubMed Links

Gene Details

Gene
CD1B
Gene Name
CD1b molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001764.2 582 Intron NP_001755.1
XM_011510119.2 582 Intron XP_011508421.1
XM_017002784.1 582 Intron XP_016858273.1
XM_017002785.1 582 Intron XP_016858274.1
XM_017002786.1 582 Intron XP_016858275.1
Gene
CD1C
Gene Name
CD1c molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001765.2 582 Missense Mutation CAG,CGG Q97R NP_001756.2
XM_005245579.4 582 Missense Mutation CAG,CGG Q97R XP_005245636.1

View Full Product Details