Product Details

SNP ID
rs114646292
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:231163875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACGGCCCGCAAGTTCCCCAC[A/G]TGTCCCGAGCATGAAATGGAGAACT
Phenotype
MIM: 610584
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM67 PubMed Links

Gene Details

Gene
TRIM67
Gene Name
tripartite motif containing 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004342.3 762 Silent Mutation ACA,ACG T302T NP_001004342.3
NM_001300889.1 762 Silent Mutation ACA,ACG T240T NP_001287818.1
XM_011544192.2 762 Silent Mutation ACA,ACG T302T XP_011542494.1
XM_017001323.1 762 Silent Mutation ACA,ACG T302T XP_016856812.1

View Full Product Details