Product Details

SNP ID
rs114968391
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:166913286 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGACACAGAAAAAAAAAGTCATG[A/G]ACAACAGTAATACAAACATTTCAGT
Phenotype
MIM: 611368
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ILDR2 PubMed Links

Gene Details

Gene
ILDR2
Gene Name
immunoglobulin like domain containing receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199351.2 7836 UTR 3 NP_955383.1
XM_006711314.3 7836 UTR 3 XP_006711377.1
XM_017001252.1 7836 UTR 3 XP_016856741.1
XM_017001253.1 7836 UTR 3 XP_016856742.1
XM_017001254.1 7836 UTR 3 XP_016856743.1
XM_017001255.1 7836 UTR 3 XP_016856744.1
XM_017001256.1 7836 UTR 3 XP_016856745.1
XM_017001257.1 7836 Intron XP_016856746.1
XM_017001258.1 7836 Intron XP_016856747.1
Gene
MAEL
Gene Name
maelstrom spermatogenic transposon silencer
There are no transcripts associated with this gene.

View Full Product Details