Product Details

SNP ID
rs115333920
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:236398931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTAACACTCAGCATAGTACTCC[A/G]CACATAGTAAGCCCTGGGCAAATAC
Phenotype
MIM: 606603
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDARADD PubMed Links

Gene Details

Gene
EDARADD
Gene Name
EDAR associated death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080738.3 Intron NP_542776.1
NM_145861.2 Intron NP_665860.2

View Full Product Details