Product Details

SNP ID
rs116022586
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:158291396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACCATGCAAGTCAAGATTACTC[A/G]AAATGTAAGTTCAATCATCTAAATT
Phenotype
MIM: 188360 MIM: 188340
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD1B PubMed Links

Gene Details

Gene
CD1B
Gene Name
CD1b molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001764.2 616 Intron NP_001755.1
XM_011510119.2 616 Intron XP_011508421.1
XM_017002784.1 616 Intron XP_016858273.1
XM_017002785.1 616 Intron XP_016858274.1
XM_017002786.1 616 Intron XP_016858275.1
Gene
CD1C
Gene Name
CD1c molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001765.2 616 Silent Mutation TCA,TCG S108S NP_001756.2
XM_005245579.4 616 Silent Mutation TCA,TCG S108S XP_005245636.1

View Full Product Details