Product Details

SNP ID
rs116467365
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:228153177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACAGTGCCCACCTCAGCTTGTCC[C/T]GCCTCTCTATCATGGTCCTTTCTTT
Phenotype
MIM: 608803 MIM: 139270
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJC2 PubMed Links

Gene Details

Gene
GJC2
Gene Name
gap junction protein gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020435.3 Intron NP_065168.2
Gene
GUK1
Gene Name
guanylate kinase 1
There are no transcripts associated with this gene.

View Full Product Details