Product Details

SNP ID
rs137965913
Assay Type
Functionally Tested
NCBI dbSNP Submissions
7
Location
Chr.1:234634584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGAAGAGTCATAATGAACTGTGC[C/T]GTCATCTACAAAGGAAGTGCAGCAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LINC00184 PubMed Links

Gene Details

Gene
LINC00184
Gene Name
long intergenic non-protein coding RNA 184
There are no transcripts associated with this gene.

View Full Product Details