Product Details

SNP ID
rs138237342
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:29148709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTGGGAGCGGTCAGTGTCTTCCT[C/T]CTTCTTCTTCTTGCTGCTGCCCGCC
Phenotype
MIM: 601940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRSF4 PubMed Links

Gene Details

Gene
SRSF4
Gene Name
serine and arginine rich splicing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005626.4 1269 Missense Mutation AAG,GAG K396E NP_005617.2
XM_011541951.2 1269 Missense Mutation AAG,GAG K366E XP_011540253.1

View Full Product Details