Product Details

SNP ID
rs138509537
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:155899009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACGTTTCTAGTCAACACAAACATT[A/G]CTTCAGTGACACTTAATATTCAATA
Phenotype
MIM: 609591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIT1 PubMed Links

Gene Details

Gene
RIT1
Gene Name
Ras like without CAAX 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256820.1 2242 UTR 3 NP_001243749.1
NM_001256821.1 2242 UTR 3 NP_001243750.1
NM_006912.5 2242 UTR 3 NP_008843.1

View Full Product Details