Product Details

SNP ID
rs139593918
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:156244737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCATCTGCCCCTGCCTCTTCCC[A/G]GGCCGGGCCAGGCGTGCCCTCACCG
Phenotype
MIM: 112260 MIM: 614579 MIM: 609176 MIM: 610962
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BGLAP PubMed Links

Gene Details

Gene
BGLAP
Gene Name
bone gamma-carboxyglutamate protein
There are no transcripts associated with this gene.

Gene
PAQR6
Gene Name
progestin and adipoQ receptor family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272104.1 907 Intron NP_001259033.1
NM_001272105.1 907 Intron NP_001259034.1
NM_001272106.1 907 Missense Mutation NP_001259035.1
NM_001272107.1 907 Intron NP_001259036.1
NM_001272108.1 907 Intron NP_001259037.1
NM_001272109.1 907 Intron NP_001259038.1
NM_001272110.1 907 Intron NP_001259039.1
NM_001272111.1 907 Intron NP_001259040.1
NM_001272112.1 907 Intron NP_001259041.1
NM_001272113.1 907 Intron NP_001259042.1
NM_024897.3 907 Missense Mutation NP_079173.2
NM_198406.2 907 Intron NP_940798.1
XM_005245494.3 907 Intron XP_005245551.1
XM_006711546.2 907 Missense Mutation XP_006711609.1
XM_006711547.3 907 Missense Mutation XP_006711610.1
XM_006711548.3 907 Missense Mutation XP_006711611.1
XM_006711552.2 907 Missense Mutation XP_006711615.1
XM_006711553.3 907 Missense Mutation XP_006711616.1
XM_011510000.2 907 Intron XP_011508302.1
XM_017002385.1 907 Missense Mutation XP_016857874.1
Gene
PMF1
Gene Name
polyamine-modulated factor 1
There are no transcripts associated with this gene.

Gene
PMF1-BGLAP
Gene Name
PMF1-BGLAP readthrough
There are no transcripts associated with this gene.

Gene
SMG5
Gene Name
SMG5, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details