Product Details

SNP ID
rs139753069
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:151760821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTATGCAAAACACTCACCGTCA[C/G]CTCACACCAATACTCGCCCCACCGT
Phenotype
MIM: 611824 MIM: 605138 MIM: 609501
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPL9 PubMed Links

Gene Details

Gene
MRPL9
Gene Name
mitochondrial ribosomal protein L9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300733.1 1104 Missense Mutation CTG,GTG L189V NP_001287662.1
NM_031420.3 1104 Missense Mutation CTG,GTG L223V NP_113608.1
Gene
OAZ3
Gene Name
ornithine decarboxylase antizyme 3
There are no transcripts associated with this gene.

Gene
TDRKH
Gene Name
tudor and KH domain containing
There are no transcripts associated with this gene.

View Full Product Details