Product Details

SNP ID
rs140027264
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:23559226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCGATGACGCGCTGTAGGATTTC[C/T]ACCTGGCTAAGCTGAGTGCCTCTCG
Phenotype
MIM: 600277
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ID3 PubMed Links

Gene Details

Gene
ID3
Gene Name
inhibitor of DNA binding 3, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002167.4 569 Silent Mutation GTA,GTG V67V NP_002158.3

View Full Product Details