Product Details

SNP ID
rs140251959
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:230979178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGGGGTGGCAGCGACTCCTACT[G/T]CTGCCTCGGCCTCCTGCCAGTACAG
Phenotype
MIM: 611647
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARV1 PubMed Links

Gene Details

Gene
ARV1
Gene Name
ARV1 homolog, fatty acid homeostasis modulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022786.1 102 Missense Mutation GCT,TCT A25S NP_073623.1
XM_017002106.1 102 Missense Mutation GCT,TCT A25S XP_016857595.1
Gene
TTC13
Gene Name
tetratricopeptide repeat domain 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122835.2 102 Intron NP_001116307.2
NM_024525.4 102 Intron NP_078801.3
XM_005273260.1 102 Intron XP_005273317.1
XM_005273261.2 102 Intron XP_005273318.1
XM_005273262.1 102 Intron XP_005273319.1
XM_005273264.2 102 Intron XP_005273321.1
XM_006711814.2 102 Intron XP_006711877.1
XM_006711815.2 102 Intron XP_006711878.1
XM_011544275.2 102 Intron XP_011542577.1
XM_011544276.2 102 Intron XP_011542578.1
XM_017002321.1 102 Intron XP_016857810.1
XM_017002322.1 102 Intron XP_016857811.1
XM_017002323.1 102 Intron XP_016857812.1
XM_017002324.1 102 Intron XP_016857813.1
XM_017002325.1 102 Intron XP_016857814.1
XM_017002326.1 102 Intron XP_016857815.1

View Full Product Details