Product Details
- SNP ID
-
rs140318994
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:242089865 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTCTTCATCATGTTATACGTTCC[A/G]GGGATCCTTTCCGCCTGTGTCGTCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PLD5
PubMed Links
Gene Details
- Gene
- PLD5
- Gene Name
- phospholipase D family member 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001195811.1 |
1827 |
Missense Mutation |
CGG,TGG |
R472W |
NP_001182740.1 |
NM_001195812.1 |
1827 |
Missense Mutation |
CGG,TGG |
R326W |
NP_001182741.1 |
NM_001320272.1 |
1827 |
Missense Mutation |
CGG,TGG |
R442W |
NP_001307201.1 |
NM_152666.2 |
1827 |
Missense Mutation |
CGG,TGG |
R534W |
NP_689879.2 |
XM_006711752.2 |
1827 |
Missense Mutation |
CGG,TGG |
R534W |
XP_006711815.1 |
XM_011544115.2 |
1827 |
Missense Mutation |
CGG,TGG |
R443W |
XP_011542417.1 |
XM_011544116.2 |
1827 |
Missense Mutation |
CGG,TGG |
R443W |
XP_011542418.1 |
XM_011544119.2 |
1827 |
Missense Mutation |
CGG,TGG |
R399W |
XP_011542421.2 |
XM_011544120.2 |
1827 |
Missense Mutation |
CGG,TGG |
R326W |
XP_011542422.1 |
XM_011544121.2 |
1827 |
Missense Mutation |
CGG,TGG |
R326W |
XP_011542423.1 |
XM_011544122.2 |
1827 |
Missense Mutation |
CGG,TGG |
R326W |
XP_011542424.1 |
XM_017000567.1 |
1827 |
Missense Mutation |
CGG,TGG |
R442W |
XP_016856056.1 |
XM_017000568.1 |
1827 |
Missense Mutation |
CGG,TGG |
R442W |
XP_016856057.1 |
XM_017000569.1 |
1827 |
Missense Mutation |
CGG,TGG |
R326W |
XP_016856058.1 |
XM_017000570.1 |
1827 |
Missense Mutation |
CGG,TGG |
R326W |
XP_016856059.1 |
View Full Product Details