Product Details

SNP ID
rs140666123
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:113980384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGTCGGCATGCTGCTGAGCTGC[A/G]GGACTTCAAGAACAAGATGCTGCCA
Phenotype
MIM: 608003 MIM: 610088
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIPK1 PubMed Links

Gene Details

Gene
HIPK1
Gene Name
homeodomain interacting protein kinase 1
There are no transcripts associated with this gene.

Gene
OLFML3
Gene Name
olfactomedin like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286352.1 445 Missense Mutation CAG,CGG Q36R NP_001273281.1
NM_001286353.1 445 UTR 5 NP_001273282.1
NM_020190.3 445 Missense Mutation CAG,CGG Q56R NP_064575.1
XM_017001848.1 445 Missense Mutation CAG,CGG Q36R XP_016857337.1

View Full Product Details