Product Details

SNP ID
rs140957303
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:114675992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATGTGGAAGGAAATTCTTGGAA[C/T]GGAACACATCACTAGAGGCAAGAAT
Phenotype
MIM: 102770
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMPD1 PubMed Links

Gene Details

Gene
AMPD1
Gene Name
adenosine monophosphate deaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000036.2 1547 Missense Mutation CCT,CTT P500L NP_000027.2
NM_001172626.1 1547 Missense Mutation CCT,CTT P496L NP_001166097.1
Gene
DENND2C
Gene Name
DENN domain containing 2C
There are no transcripts associated with this gene.

View Full Product Details