Product Details

SNP ID
rs141689121
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:154569502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGAGGAGCGGCTGGTGGAGCA[A/T]CTCCTGGATCCTTCCCGCTACAACA
Phenotype
MIM: 118507
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 351 Missense Mutation CAA,CAT Q35H NP_000739.1
XM_017000180.1 351 UTR 5 XP_016855669.1

View Full Product Details