Product Details

SNP ID
rs142466418
Assay Type
Functionally Tested
NCBI dbSNP Submissions
14
Location
Chr.1:23559002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGTCGTTGGAGATGACAAGTTC[C/T]GGAGTGAGCTCGGCTGTCTGATTAG
Phenotype
MIM: 600277
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ID3 PubMed Links
Additional Information
For this assay, SNP(s) [rs11574] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ID3
Gene Name
inhibitor of DNA binding 3, HLH protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002167.4 686 Silent Mutation NP_002158.3

View Full Product Details