Product Details

SNP ID
rs143155155
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:26190694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATACAGGCCTCGAGGGGTGGGGC[A/G]GGACTCAGGAGGACCGTATGGGGTT
Phenotype
MIM: 609121 MIM: 603272
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CATSPER4 PubMed Links

Gene Details

Gene
CATSPER4
Gene Name
cation channel sperm associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198137.1 67 Missense Mutation AGG,GGG R23G NP_937770.1
XM_011541432.2 67 Missense Mutation AGG,GGG R23G XP_011539734.1
XM_011541433.2 67 Missense Mutation AGG,GGG R23G XP_011539735.1
Gene
CNKSR1
Gene Name
connector enhancer of kinase suppressor of Ras 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297647.1 67 Intron NP_001284576.1
NM_001297648.1 67 Intron NP_001284577.1
NM_006314.2 67 Intron NP_006305.2
XM_011540484.2 67 Intron XP_011538786.1
XM_011540485.2 67 Intron XP_011538787.1

View Full Product Details