Product Details

SNP ID
rs143245095
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:246842733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTTTGACGTCCTGATTCGTTTC[A/G]CTTTTGGCTTTCCCAGAGTTTCTAC
Phenotype
MIM: 610853
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AHCTF1 PubMed Links

Gene Details

Gene
AHCTF1
Gene Name
AT-hook containing transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323342.1 6977 Missense Mutation GCG,GTG A2190V NP_001310271.1
NM_001323343.1 6977 Missense Mutation GCG,GTG A2190V NP_001310272.1
NM_015446.4 6977 Intron NP_056261.4
XM_006711758.1 6977 Intron XP_006711821.1
XM_011544156.2 6977 Intron XP_011542458.2
XM_011544157.2 6977 Intron XP_011542459.2

View Full Product Details