Product Details

SNP ID
rs143615282
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:182905628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCACCATGCTGTTGGAGAAGTG[A/T]GTCTGAAGATAAATCCTTTATCTAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SHCBP1L PubMed Links

Gene Details

Gene
SHCBP1L
Gene Name
SHC binding and spindle associated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030933.2 1489 Missense Mutation ACA,TCA T402S NP_112195.2

View Full Product Details