Product Details

SNP ID
rs144361455
Assay Type
Functionally Tested
NCBI dbSNP Submissions
7
Location
Chr.1:26190752 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTAGCTGCACTGAGGGGCCGCC[C/G]CTCTCCCCTGCAGAGTACCATTCAC
Phenotype
MIM: 609121 MIM: 603272
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
CATSPER4 PubMed Links
Additional Information
For this assay, SNP(s) [rs41284333] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CATSPER4
Gene Name
cation channel sperm associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198137.1 125 Missense Mutation CCC,CGC P42R NP_937770.1
XM_011541432.2 125 Missense Mutation CCC,CGC P42R XP_011539734.1
XM_011541433.2 125 Missense Mutation CCC,CGC P42R XP_011539735.1
Gene
CNKSR1
Gene Name
connector enhancer of kinase suppressor of Ras 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297647.1 125 Intron NP_001284576.1
NM_001297648.1 125 Intron NP_001284577.1
NM_006314.2 125 Intron NP_006305.2
XM_011540484.2 125 Intron XP_011538786.1
XM_011540485.2 125 Intron XP_011538787.1

View Full Product Details