Product Details

SNP ID
rs144441230
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:168246033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTCTCAATGGACGCAGTGACGC[A/G]ATGAAGCATCCAGCAAAGCTTTTGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD36BP1 PubMed Links

Gene Details

Gene
ANKRD36BP1
Gene Name
ankyrin repeat domain 36B pseudogene 1
There are no transcripts associated with this gene.

Gene
SFT2D2
Gene Name
SFT2 domain containing 2
There are no transcripts associated with this gene.

View Full Product Details