Product Details

SNP ID
rs144570390
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:182905532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCATCTGTCAACAAAGCAAGAT[G/T]TGCAGCTTGATATTCTCCTGGAAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SHCBP1L PubMed Links

Gene Details

Gene
SHCBP1L
Gene Name
SHC binding and spindle associated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030933.2 1585 Missense Mutation AAT,CAT N434H NP_112195.2

View Full Product Details