Product Details

SNP ID
rs145225693
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:226225538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCAGAACAGGCGTGCCAAGTCTC[G/T]GCGTCAGAGTGGGAAATCCTTCCAA
Phenotype
MIM: 609375 MIM: 609852
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LIN9 PubMed Links

Gene Details

Gene
LIN9
Gene Name
lin-9 DREAM MuvB core complex component
There are no transcripts associated with this gene.

Gene
MIXL1
Gene Name
Mix paired-like homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282402.1 513 Missense Mutation CGG,CTG R150L NP_001269331.1
NM_031944.2 513 Missense Mutation CGG,CTG R142L NP_114150.1

View Full Product Details