Product Details

SNP ID
rs145239312
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:182901521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGAACAAGGTTAATAATTTCTG[C/G]GAACACAAACAAAGCCAGTGTGGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SHCBP1L PubMed Links

Gene Details

Gene
SHCBP1L
Gene Name
SHC binding and spindle associated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030933.2 Intron NP_112195.2

View Full Product Details