Product Details
- SNP ID
-
rs146360764
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:111414627 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTTCCGGGAGCTGGATGACGGGG[A/C]TGGAGGACAGCATCATCCTGTTTTC
- Phenotype
-
MIM: 603578
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OVGP1
PubMed Links
Gene Details
- Gene
- OVGP1
- Gene Name
- oviductal glycoprotein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002557.3 |
1930 |
Missense Mutation |
AGC,ATC |
S625I |
NP_002548.3 |
View Full Product Details