Product Details
- SNP ID
-
rs146585589
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:29148796 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTCTTGCTGCGGCTGCGACTGC[A/G]ACTGCGGCTCTCCTCTCTGCTTCTC
- Phenotype
-
MIM: 601940
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SRSF4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2230678] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SRSF4
- Gene Name
- serine and arginine rich splicing factor 4
View Full Product Details