Product Details

SNP ID
rs146741741
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:3489110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACACTCCGTCTGCACCCCAGTCC[A/G]TCTCTCTCTGGCTGCTGCTTCTGCA
Phenotype
MIM: 604266
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF16 PubMed Links

Gene Details

Gene
ARHGEF16
Gene Name
Rho guanine nucleotide exchange factor 16
There are no transcripts associated with this gene.

Gene
MEGF6
Gene Name
multiple EGF like domains 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001409.3 5906 UTR 3 NP_001400.3
XM_006710406.3 5906 UTR 3 XP_006710469.1
XM_011540885.1 5906 UTR 3 XP_011539187.1
XM_011540886.1 5906 UTR 3 XP_011539188.1
XM_011540887.2 5906 UTR 3 XP_011539189.1
XM_011540888.2 5906 Intron XP_011539190.1
XM_017000533.1 5906 UTR 3 XP_016856022.1

View Full Product Details