Product Details

SNP ID
rs147002798
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:26191311 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACGCCTGGGACATGCAGGAGTTC[A/G]TCACTCACATGTACATCAAGCAGCT
Phenotype
MIM: 609121 MIM: 603272
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CATSPER4 PubMed Links

Gene Details

Gene
CATSPER4
Gene Name
cation channel sperm associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198137.1 238 Missense Mutation ATC,GTC I80V NP_937770.1
XM_011541432.2 238 Missense Mutation ATC,GTC I80V XP_011539734.1
XM_011541433.2 238 Missense Mutation ATC,GTC I80V XP_011539735.1
Gene
CNKSR1
Gene Name
connector enhancer of kinase suppressor of Ras 1
There are no transcripts associated with this gene.

View Full Product Details