Product Details

SNP ID
rs147954793
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:228158061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTACCTGGGCTACGCCGTGCACCG[C/T]CTGGCCCGTGCGTCTGAGCAGGAGC
Phenotype
MIM: 608803 MIM: 139270 MIM: 615316
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJC2 PubMed Links

Gene Details

Gene
GJC2
Gene Name
gap junction protein gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020435.3 616 Silent Mutation CGC,CGT R101R NP_065168.2
Gene
GUK1
Gene Name
guanylate kinase 1
There are no transcripts associated with this gene.

Gene
IBA57
Gene Name
IBA57 homolog, iron-sulfur cluster assembly
There are no transcripts associated with this gene.

Gene
IBA57-AS1
Gene Name
IBA57 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details