Product Details

SNP ID
rs148084661
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:29148545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATTCTGATGGAAGGTTTGGTTT[C/T]GATTTGGAATTGGATCTCGACCTGG
Phenotype
MIM: 601940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRSF4 PubMed Links

Gene Details

Gene
SRSF4
Gene Name
serine and arginine rich splicing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005626.4 1433 Silent Mutation TCA,TCG S450S NP_005617.2
XM_011541951.2 1433 Silent Mutation TCA,TCG S420S XP_011540253.1

View Full Product Details