Product Details

SNP ID
rs148533601
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:182905560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTTGATATTCTCCTGGAAAAAT[C/T]ATTACTGTATCTCCACTATAACAAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHCBP1L PubMed Links

Gene Details

Gene
SHCBP1L
Gene Name
SHC binding and spindle associated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030933.2 1557 Missense Mutation ATA,ATG I424M NP_112195.2

View Full Product Details