Product Details
- SNP ID
-
rs149799938
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
15
- Location
-
Chr.1:26191302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCTTCCAGGACGCCTGGGACATG[C/T]AGGAGTTCATCACTCACATGTACAT
- Phenotype
-
MIM: 609121
MIM: 603272
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CATSPER4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11247866] are located under a probe and SNP(s) [rs11247865] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CATSPER4
- Gene Name
- cation channel sperm associated 4
- Gene
- CNKSR1
- Gene Name
- connector enhancer of kinase suppressor of Ras 1
There are no transcripts associated with this gene.
View Full Product Details