Product Details
- SNP ID
-
rs149812110
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:152910065 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCACAGGAGCAGGAGCTGCAGCAA[C/T]AGCACTGGGAACAGCATGAGGAATA
- Phenotype
-
MIM: 147360
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IVL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs200220063] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IVL
- Gene Name
- involucrin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005547.2 |
314 |
Nonsense Mutation |
CAG,TAG |
Q90* |
NP_005538.2 |
View Full Product Details