Product Details

SNP ID
rs150303136
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:23629527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGAGACTTTGTGCCATCCTCTG[C/G]TATATTATCCTGAAGAGTGCCTGGC
Phenotype
MIM: 607305
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MDS2 PubMed Links

Gene Details

Gene
MDS2
Gene Name
myelodysplastic syndrome 2 translocation associated
There are no transcripts associated with this gene.

View Full Product Details