Product Details

SNP ID
rs150305306
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:86428521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGAAAATACATTCATTTCACAC[C/T]TAATTTCCTACTGAATGATAACTTA
Phenotype
MIM: 604003
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCA2 PubMed Links

Gene Details

Gene
CLCA2
Gene Name
chloride channel accessory 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006536.5 590 Missense Mutation CCT,CTT P143L NP_006527.1
XM_011542448.2 590 Missense Mutation CCT,CTT P143L XP_011540750.1

View Full Product Details