Product Details

SNP ID
rs150761078
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:53461602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTGGCGTCCCCCTGCAGCAGG[A/G]CTTCCGGCATGTGTCCCGCAGCCAG
Phenotype
MIM: 614805
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMRTB1 PubMed Links

Gene Details

Gene
DMRTB1
Gene Name
DMRT like family B with proline rich C-terminal 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033067.2 762 Missense Mutation GAC,GGC D236G NP_149056.1

View Full Product Details