Product Details

SNP ID
rs150827657
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:235448335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGAGAGAACGAATGGACTTTTC[C/T]TGTCTTTTGATAGGCTCCATGACAA
Phenotype
MIM: 610194 MIM: 604934
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GALNT2 PubMed Links

Gene Details

Gene
B3GALNT2
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277155.2 3690 Intron NP_001264084.1
NM_152490.4 3690 UTR 3 NP_689703.1
XM_006711749.3 3690 Intron XP_006711812.1
XM_017000394.1 3690 UTR 3 XP_016855883.1
XM_017000395.1 3690 UTR 3 XP_016855884.1
Gene
TBCE
Gene Name
tubulin folding cofactor E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079515.2 3690 Intron NP_001072983.1
NM_001287801.1 3690 Intron NP_001274730.1
NM_001287802.1 3690 Intron NP_001274731.1
NM_003193.4 3690 Intron NP_003184.1

View Full Product Details